Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSPH1
Entrez Gene ID
102173879
Gene Name
binder of sperm protein homolog 1
Gene Aliases
-
Chromosome Location
Chr.18: 55854470 - 55862059 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971381.1 XP_013826835.1 3 - 4 230 90

View Full Product Details