Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM31
Entrez Gene ID
102173141
Gene Name
tripartite motif containing 31
Gene Aliases
-
Chromosome Location
Chr.23: 21193178 - 21222416 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038813.1 XP_017894302.1 4 - 5 1230 74

View Full Product Details