Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH4
Entrez Gene ID
102186165
Gene Name
membrane associated ring-CH-type finger 4
Gene Aliases
-
Chromosome Location
Chr.2: 31421704 - 31541806 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676518.3 XP_005676575.2 1 - 2 2611 84

View Full Product Details