Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNDIG1L
Entrez Gene ID
102189764
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
-
Chromosome Location
Chr.10: 17393009 - 17410762 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686075.3 XP_005686132.1 3 - 4 977 77
GenBank mRNA JO644903.2 - 2 - 3 309 77

View Full Product Details