Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS30
Entrez Gene ID
102187267
Gene Name
mitochondrial ribosomal protein S30
Gene Aliases
-
Chromosome Location
Chr.20: 30046942 - 30055463 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065760.1 XP_017921249.1 2 - 3 789 96
GenBank mRNA JO434749.2 - 2 - 3 763 96

View Full Product Details