Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102188106
Entrez Gene ID
102188106
Gene Name
lipase member J
Gene Aliases
LIPJ
Chromosome Location
Chr.26: 41209421 - 41230805 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698181.2 XP_005698238.2 2 - 3 327 86

View Full Product Details