Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIC6
Entrez Gene ID
102190458
Gene Name
chloride intracellular channel 6
Gene Aliases
-
Chromosome Location
Chr.1: 41052571 - 41113308 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964117.2 XP_013819571.2 2 - 3 1382 84

View Full Product Details