Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GGT1
Entrez Gene ID
102182996
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017190051: 88409 - 105708 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045029.1 XP_017900518.1 10 - 11 2244 62
GenBank mRNA JO425797.1 - 10 - 11 1450 62

View Full Product Details