Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK3
Entrez Gene ID
102189157
Gene Name
cyclin dependent kinase 3
Gene Aliases
-
Chromosome Location
Chr.19: 54869266 - 54873100 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972144.2 XP_013827598.1 7 - 8 1005 72
XM_013972145.2 XP_013827599.1 6 - 7 938 72
XM_018063772.1 XP_017919261.1 7 - 8 1002 72
XM_018063773.1 XP_017919262.1 7 - 8 990 72
GenBank mRNA JO504710.2 - 2 - 3 724 72
KA336021.1 - 1 - 2 230 72

View Full Product Details