Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIR1
Entrez Gene ID
102183110
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
-
Chromosome Location
Chr.2: 113871332 - 113908015 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675946.3 XP_005676003.2 1 - 2 165 144

View Full Product Details