Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANAPC10
Entrez Gene ID
102177553
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
-
Chromosome Location
Chr.17: 57695395 - 57781202 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691210.3 XP_005691267.3 3 - 4 913 104
XM_018061572.1 XP_017917061.1 3 - 4 485 104
XM_018061573.1 XP_017917062.1 3 - 4 322 104
GenBank mRNA KA308255.1 - 2 - 3 434 104

View Full Product Details