Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CES3
Entrez Gene ID
102177362
Gene Name
carboxylesterase 3
Gene Aliases
-
Chromosome Location
Chr.18: 35924647 - 35937281 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692115.3 XP_005692172.2 9 - 10 1196 81
GenBank mRNA JO419999.2 - 9 - 10 1260 81
JO431746.2 - 9 - 10 1217 81

View Full Product Details