The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018062087.1 | XP_017917576.1 | 2 - 3 | 631 | 66 |
GenBank mRNA | JO426150.1 | - | 2 - 3 | 635 | 66 |