Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP35
Entrez Gene ID
102181927
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
Chromosome Location
Chr.29: 17260318 - 17326281 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA324011.1 - 1 - 2 1013 95

View Full Product Details