Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A13
Entrez Gene ID
102172466
Gene Name
membrane spanning 4-domains A13
Gene Aliases
-
Chromosome Location
Chr.15: 162113 - 183808 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059012.1 XP_017914501.1 1 - 2 141 98

View Full Product Details