Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM270
Entrez Gene ID
102177951
Gene Name
transmembrane protein 270
Gene Aliases
WBSCR28
Chromosome Location
Chr.25: 33247510 - 33249094 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040157.1 XP_017895646.1 1 - 2 96 53

View Full Product Details