Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DEGS2
Entrez Gene ID
102179052
Gene Name
delta 4-desaturase, sphingolipid 2
Gene Aliases
-
Chromosome Location
Chr.21: 64368663 - 64388056 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066041.1 XP_017921530.1 2 - 3 908 65
XM_018066042.1 XP_017921531.1 3 - 4 1026 65
GenBank mRNA JO427947.1 - 1 - 2 429 65

View Full Product Details