Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPLX2
Entrez Gene ID
102191617
Gene Name
complexin 2
Gene Aliases
-
Chromosome Location
Chr.10: 95863370 - 95952593 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053709.1 XP_017909198.1 3 - 4 589 57
XM_018053710.1 XP_017909199.1 3 - 4 579 57
XM_018053711.1 XP_017909200.1 3 - 4 615 57
GenBank mRNA JO632712.2 - 1 - 2 4106 57

View Full Product Details