Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT61B
Entrez Gene ID
102170526
Gene Name
tRNA methyltransferase 61B
Gene Aliases
-
Chromosome Location
Chr.11: 70658312 - 70671235 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686858.3 XP_005686915.1 6 - 7 2068 89
XM_005686859.3 XP_005686916.1 6 - 7 2040 89

View Full Product Details