Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LIN7B
Entrez Gene ID
102181892
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Gene Aliases
-
Chromosome Location
Chr.18: 56641126 - 56644771 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062760.1 XP_017918249.1 3 - 4 295 79

View Full Product Details