Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C20H5orf22
Entrez Gene ID
102181814
Gene Name
chromosome 20 C5orf22 homolog
Gene Aliases
-
Chromosome Location
Chr.20: 41908820 - 41930716 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694829.3 XP_005694886.1 5 - 6 951 111
GenBank mRNA JO437753.2 - 4 - 5 1157 111
JO589490.2 - 2 - 3 80 111

View Full Product Details