Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IP6K1
Entrez Gene ID
102178224
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
-
Chromosome Location
Chr.22: 50518404 - 50556353 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695950.3 XP_005696007.2 5 - 6 1439 81
XM_018066660.1 XP_017922149.1 5 - 6 1141 81
XM_018066661.1 XP_017922150.1 4 - 5 1088 81
XM_018066662.1 XP_017922151.1 4 - 5 755 81
GenBank mRNA JO576696.1 - 1 - 2 266 81

View Full Product Details