Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ZFP91
Entrez Gene ID
102190153
Gene Name
ZFP91 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.15: 1451745 - 1488691 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059035.1 XP_017914524.1 3 - 4 583 68
GenBank mRNA JO430053.2 - 3 - 4 394 68
JO632649.2 - 3 - 4 737 68

View Full Product Details