Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPA33
Entrez Gene ID
102190184
Gene Name
glycoprotein A33
Gene Aliases
-
Chromosome Location
Chr.3: 118354052 - 118405566 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046605.1 XP_017902094.1 5 - 6 764 62

View Full Product Details