Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MANSC4
Entrez Gene ID
102189743
Gene Name
MANSC domain containing 4
Gene Aliases
-
Chromosome Location
Chr.5: 80646965 - 80660800 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680717.3 XP_005680774.1 3 - 4 1443 63

View Full Product Details