Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GML
Entrez Gene ID
102174759
Gene Name
glycosylphosphatidylinositol anchored molecule like
Gene Aliases
-
Chromosome Location
Chr.14: 80477912 - 80508900 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058130.1 XP_017913619.1 3 - 4 275 101

View Full Product Details