Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGBL1
Entrez Gene ID
102185399
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
-
Chromosome Location
Chr.21: 16295915 - 17139898 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066611.1 XP_017922100.1 5 - 6 613 66

View Full Product Details