Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP161
Entrez Gene ID
102175992
Gene Name
cilia and flagella associated protein 161
Gene Aliases
-
Chromosome Location
Chr.21: 26575503 - 26596095 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695072.3 XP_005695129.1 5 - 6 676 80

View Full Product Details