Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPTXR
Entrez Gene ID
102183945
Gene Name
neuronal pentraxin receptor
Gene Aliases
-
Chromosome Location
Chr.5: 109346353 - 109368575 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049072.1 XP_017904561.1 3 - 4 1440 78

View Full Product Details