Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1GALT1
Entrez Gene ID
102169248
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.4: 105345142 - 105396206 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047386.1 XP_017902875.1 3 - 4 1122 100
XM_018047387.1 XP_017902876.1 4 - 5 1399 100
XM_018047389.1 XP_017902878.1 3 - 4 974 100

View Full Product Details