Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERTAD4
Entrez Gene ID
102189231
Gene Name
SERTA domain containing 4
Gene Aliases
-
Chromosome Location
Chr.16: 71719131 - 71733080 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691031.3 XP_005691088.1 3 - 4 326 112
XM_018060772.1 XP_017916261.1 3 - 4 714 112

View Full Product Details