Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM32A
Entrez Gene ID
106502278
Gene Name
family with sequence similarity 32 member A
Gene Aliases
-
Chromosome Location
Chr.7: 101929611 - 101935724 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013965028.2 XP_013820482.1 3 - 4 350 66

View Full Product Details