Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRT3
Entrez Gene ID
102183189
Gene Name
proline rich transmembrane protein 3
Gene Aliases
-
Chromosome Location
Chr.22: 16637742 - 16644600 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066829.1 XP_017922318.1 2 - 3 1198 86
GenBank mRNA JO618960.2 - 2 - 3 501 86

View Full Product Details