Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2H2orf76
Entrez Gene ID
102182546
Gene Name
chromosome 2 C2orf76 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 64937332 - 65032602 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO619775.2 - 2 - 3 202 104

View Full Product Details