Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C25H16orf58
Entrez Gene ID
102181919
Gene Name
chromosome 25 C16orf58 homolog
Gene Aliases
-
Chromosome Location
Chr.25: 27398124 - 27413878 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040643.1 XP_017896132.1 7 - 8 784 79
GenBank mRNA JO637366.2 - 7 - 8 744 79

View Full Product Details