Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A34
Entrez Gene ID
102173215
Gene Name
solute carrier family 25 member 34
Gene Aliases
-
Chromosome Location
Chr.16: 50309455 - 50315161 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013970278.2 XP_013825732.2 4 - 5 2235 93

View Full Product Details