Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSGIN1
Entrez Gene ID
102181147
Gene Name
oxidative stress induced growth inhibitor 1
Gene Aliases
-
Chromosome Location
Chr.18: 11677729 - 11688696 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061773.1 XP_017917262.1 2 - 3 326 76
XM_018061774.1 XP_017917263.1 2 - 3 260 76
GenBank mRNA JO412150.2 - 2 - 3 238 76

View Full Product Details