Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D1A
Entrez Gene ID
102171584
Gene Name
SH2 domain containing 1A
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 17202278 - 17223238 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700359.3 XP_005700416.1 3 - 4 515 86

View Full Product Details