Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NYX
Entrez Gene ID
102180385
Gene Name
nyctalopin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189629: 314806 - 334751 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044569.1 XP_017900058.1 2 - 2 1421 82

View Full Product Details