Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP8
Entrez Gene ID
102171586
Gene Name
Rho GTPase activating protein 8
Gene Aliases
-
Chromosome Location
Chr.5: 114253571 - 114307346 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048888.1 XP_017904377.1 11 - 12 1139 91
XM_018048889.1 XP_017904378.1 10 - 11 1007 91

View Full Product Details