Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN19
Entrez Gene ID
102191306
Gene Name
tetraspanin 19
Gene Aliases
-
Chromosome Location
Chr.5: 14499634 - 14530452 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679776.2 XP_005679833.2 3 - 4 217 112
XM_018049084.1 XP_017904573.1 3 - 4 215 112

View Full Product Details