Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZKSCAN5
Entrez Gene ID
102187285
Gene Name
zinc finger with KRAB and SCAN domains 5
Gene Aliases
-
Chromosome Location
Chr.25: 37779158 - 37796914 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697844.3 XP_005697901.1 5 - 6 1415 75
XM_005697845.3 XP_005697902.1 5 - 6 1248 75
XM_005697846.3 XP_005697903.1 5 - 6 933 75
XM_005697847.3 XP_005697904.1 5 - 6 925 75
XM_005697848.3 XP_005697905.1 5 - 6 1412 75
XM_005697850.3 XP_005697907.1 5 - 6 1413 75
XM_013974681.2 XP_013830135.1 5 - 6 918 75
XM_013974683.2 XP_013830137.1 5 - 6 1245 75
XM_018039873.1 XP_017895362.1 6 - 7 1570 75

View Full Product Details