Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PGLYRP1
Entrez Gene ID
102168570
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 54562176 - 54566407 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062639.1 XP_017918128.1 2 - 3 522 70
GenBank mRNA JO340320.2 - 2 - 3 424 70
KA340135.1 - 2 - 3 535 70

View Full Product Details