Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC108634317
Entrez Gene ID
108634317
Gene Name
uncharacterized protein CXorf49 homolog
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 10190998 - 10195134 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043721.1 XP_017899210.1 4 - 5 1708 88

View Full Product Details