Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC8
Entrez Gene ID
102189881
Gene Name
WAP four-disulfide core domain 8
Gene Aliases
-
Chromosome Location
Chr.13: 73910719 - 73943336 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056874.1 XP_017912363.1 5 - 6 634 57

View Full Product Details