Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTUB2
Entrez Gene ID
102184847
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Gene Aliases
-
Chromosome Location
Chr.21: 58072020 - 58093228 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066511.1 XP_017922000.1 3 - 4 425 81
GenBank mRNA KA332420.1 - 3 - 4 3267 81

View Full Product Details