Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102183400
Entrez Gene ID
102183400
Gene Name
nuclear RNA export factor 2
Gene Aliases
NXF2B
Chromosome Location
Chr.X|NW_017189517: 41207265 - 41223686 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044110.1 XP_017899599.1 21 - 22 1918 63

View Full Product Details