Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC66
Entrez Gene ID
102168990
Gene Name
leucine rich repeat containing 66
Gene Aliases
-
Chromosome Location
Chr.6: 68373503 - 68404240 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681606.3 XP_005681663.3 3 - 4 947 65
XM_018049451.1 XP_017904940.1 3 - 4 1022 65
GenBank mRNA JT663224.1 - 5 - 6 2014 65

View Full Product Details