Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL50
Entrez Gene ID
102174845
Gene Name
mitochondrial ribosomal protein L50
Gene Aliases
-
Chromosome Location
Chr.8: 90911309 - 90918860 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684246.3 XP_005684303.1 1 - 2 128 67

View Full Product Details