Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS21
Entrez Gene ID
102169499
Gene Name
regulator of G protein signaling 21
Gene Aliases
-
Chromosome Location
Chr.16: 11498039 - 11523790 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690463.2 XP_005690520.2 2 - 3 255 83

View Full Product Details